Order Entry
Austria
ContactUsLinkComponent
ASTM® D2887/D5307 column resolution Test Mix, certified reference material, Supelco®
ASTM® D2887/D5307 column resolution Test Mix, certified reference material, Supelco®
Catalog # 48889.
Supplier:  Merck
CAS Number:  
ASTM® D2887/D5307 column resolution Test Mix, certified reference material, Supelco®
Catalog # 48889.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 % (w/v) each component in octane
  • Pk:
    1
  • Description:
    ASTM(R) D2887/D5307 Column Resolution Test Mix

Specifications

About this item