Order Entry
Austria
ContactUsLinkComponent
ASTM® 5442 (C12-C60) quantitative linearity standard, analytical standard, Supelco®
ASTM® 5442 (C12-C60) quantitative linearity standard, analytical standard, Supelco®
Catalog # 502235.
Supplier:  Merck
CAS Number:  
ASTM® 5442 (C12-C60) quantitative linearity standard, analytical standard, Supelco®
Catalog # 502235.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.01 % (w/w) each component in cyclohexane
  • Pk:
    1
  • Description:
    ASTM(R) 5442 (C12-C60) Quantitative Linearity Standard

Specifications

About this item