Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Alkane Standard Solution C₂₁-C₄₀
Supelco® Organic Reference Standard, Alkane Standard Solution C₂₁-C₄₀
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Alkane Standard Solution C₂₁-C₄₀
Supplier:  Merck

Specifications

  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H361d - H373 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –4 °C - closed cup

Specifications

Frequently Bought Together