Order Entry
Austria
ContactUsLinkComponent
Spurr low viscosity embedding kit, Sigma-Aldrich®
Spurr low viscosity embedding kit, Sigma-Aldrich®
Catalog # EM0300-1KT
CAS Number:  
Spurr low viscosity embedding kit, Sigma-Aldrich®
Catalog # EM0300-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Spurr Low Viscosity Embedding Kit, Sigma-Aldrich®

Specifications

About this item

Spurr resin is a hydrophobic resin. It is less viscous than epon mixtures and is easier to dispense. The low viscosity of the resin contributes to its penetration in tissue, mineral and dense structures. A low viscosity of 60 cps also allows easy penetration into a variety of difficult materials.

  • Quality Level 200

Exceptional penetration for tissue, mineral, and dense structures. A low viscosity of 60 cps allows easy penetration into a variety of difficult materials. Spurr low viscosity embedding kit has been used to embed histological samples like tissues, organs and embryos.[2][3][4] It may be used to prepare mineral specimens for polishing. The low viscosity of the Spurr formulation allows rapid infiltration into tissues, minerals, and other dense structures.

Kit contains:

Nonenylsuccinic anhydride 450 g
D.E.R. 736 250 g
Dimethylaminoethanol 100 g
ERL 4221 250 g