Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Decalcifying Solution-Lite in aqueous solution

Glass bottle, 1 L
Catalog # D0818-1L
Corrosive
Decalcifying Solution-Lite in aqueous solution
Catalog # D0818-1L
CAS Number:  
Decalcifying Solution-Lite in aqueous solution

Glass bottle, 1 L

CAS Number:  
Price
58,60 €
Log in for availability
1 * 1 L
Product Details & Documents

Hydrochloric acid based aqueous solution. Decalcifying Solution-Lite is designed to be a universal effective decalcifying agent. It is intended to be used for the decalcification of routine, immunohistochemical and bone marrow core specimens.

Decalcifying Solution-Lite has been tailored to suit your specific lab routine. It works equally well with all types of specimens in an easy to handle time frame.

  • Boiling Pt:208 °F
  • Storage Temperature:Ambient
  • UN:1789
  • ADR:8,III
Specifications

Specification Test Results

  • Appearance:
    Yellow to amber water like liquid with suspended dark particulate that will accumulate and intesify over time.

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications