Order Entry
Austria
ContactUsLinkComponent
Automated cell counter, EVE™ Plus
Automated cell counter, EVE™ Plus
Catalog # 734-3333
Supplier:  NANOENTEK
undefined
Automated cell counter, EVE™ Plus
Catalog # 734-3333
Supplier:  NANOENTEK

Specifications

  • Model:
    EVE™ Plus
  • Sample volume:
    10 µl
  • Description:
    Automated cell counter, EU-plug
  • Seq:
    02
  • Cell size:
    5 - 60 µm
  • Concentration:
    1×10⁴ - 2×10⁷ cells/ml
  • Focus:
    Manual or automatic
  • Measurement time:
    1 s/test (manual focus), <10 s/test (auto focus)
  • Pk:
    1

Specifications

About this item

EVE™ Plus is a compact image-based automated cell counter that uses disposable slides to most accurately determine eukaryotic cell titers in less than one second. In addition, when applying trypan blue staining, the system will discriminate between live from dead cells precisely to provide reliable data on cell viability. Compared to counting cells manually using classical haemocytometers, the EVE™ PLUS automated cell counter offers three main benefits.

  • Reproducible, objective data, free from human bias/error
  • Quick and lean workflow saves times and increases productivity
  • Disposable counting slides improve lab safety

Based on its well established precursor Eve™, the EVE™ Plus uses advanced optics and image analysis algorithms for counting, even challenging clumped cell populations, at a precision that is proven to be comparable to more complex fluorescence-based counting methods.
The cell concentration measuring range has been increased to 1×10⁴ to 2×10⁷ cells/ml with cell sizes ranging from 5 to 60 µm being recognised. The system can, therefore, be used on all kinds of round shaped mammalian cell lines, primary cells, red blood cells, PBMC, and some round types of yeast. For irregular shaped cells we recommend you use the ADAM-MC2 fluorescence-based cell counter.

A well-arranged touch screen user interface together with smart data storage and transfer options guarantee for easy and hassle-free operation.

EVE cell counting slides offer reproducible and reliable measurements and analysis. The unique design eliminates the use of cover slips and allows for exact volume control (10 µl cell suspension per test). In contrast to classical haemocytometers the disposable slides reduce the user’s risk of exposure to biological samples and eliminate the need for washing or autoclaving components. Each slide contains two enclosed chambers allowing for two tests per slide.

To prevent biased, unreliable results when using poor quality replicas, EVE slide packs do include an individual access code, which you must enter in the EVE PLUS software for authentication. Each access code is valid for up to 200 tests and can be used on an unlimited number of EVE PLUS instruments.