Order Entry
Austria
ContactUsLinkComponent
Tetrabutylammonium hydrogen sulphate concentrate for ion pair chromatography, Supelco®
Tetrabutylammonium hydrogen sulphate concentrate for ion pair chromatography, Supelco®
Catalog # 98479-6X1AMP-F
Supplier:  Merck
CAS Number:  
Tetrabutylammonium hydrogen sulphate concentrate for ion pair chromatography, Supelco®
Catalog # 98479-6X1AMP-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    6
  • Seq:
    0006
  • Pk Info:
    6× 1 ampoule

Specifications

About this item

 :
 :339.54 g/mol
CAS Number:32503-27-8
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637