Order Entry
Austria
ContactUsLinkComponent
 
Flammable
Corrosive
Irritant
Bis(trimethylsilyl)acetamide + Trimethylchlorosilane, (BSA+TMCS) BSA 93 - 97%, LiChropur™ for GC derivatisation, Supelco®
Catalog # 15256-10ML
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Irritant
Bis(trimethylsilyl)acetamide + Trimethylchlorosilane, (BSA+TMCS) BSA 93 - 97%, LiChropur™ for GC derivatisation, Supelco®
Catalog # 15256-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275
  • Pk Info:
    1× 10 ml

Specifications

About this item

 :
ADR:8(3),II
 :M-Clarity™ quality level = MQ100
UN:2924
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
 :10416-59-8