Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Turbidity Standards
Calibration Cylinder Kit, for Models 1720 E, D and C
Calibration Cylinder Kit, for Models 1720 E, D and C
Calibration Cylinder Kit, for Models 1720 E, D and C
Catalog # 708-0006
Supplier:
Hach
CAS Number:
Expand image
Calibration Cylinder Kit, for Models 1720 E, D and C
Catalog # 708-0006
Supplier:
Hach
CAS Number:
Specifications
Pk:
1 KIT
Turbidity:
4000 NTU
Description:
Stablcal® stabilized formazine standard
Seq:
10
Pk Info:
5×100 ml
Specifications
About this item
Product Details
Product Details