Pesticide standard, Isofenphos-oxon, reference material, Honeywell Fluka™
Catalog # HONCR1696-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:31120-85-1
- Description:Isofenphos-oxon
- Formula:C₁₅H₂₄NO₅P
- Storage temperature:2-8°C
- Description:Isofenphos-oxon
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.