Order Entry
Austria
ContactUsLinkComponent
 
Pesticide standard, Disulfoton-sulfone, reference material, Honeywell Fluka™
  HONCR1695-50MG
 :  
Pesticide standard, Disulfoton-sulfone, reference material, Honeywell Fluka™
  HONCR1695-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    2497-06-5
  • Description:
    Disulfoton-sulfone
  • Formula:
    C₈H₁₉O₄PS₃
  • Storage temperature:
    2-8°C
  • Description:
    Disulfoton-sulfone
  • Form:
    Liquid

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.