Order Entry
Austria
ContactUsLinkComponent
 
Pesticide standard, Methiocarb-sulfoxide, reference material, Honeywell Fluka™
  HONCR1694-100MG
 :  
Pesticide standard, Methiocarb-sulfoxide, reference material, Honeywell Fluka™
  HONCR1694-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    2635-10-1
  • Description:
    Methiocarb-sulfoxide
  • Formula:
    C₁₁H₁₅NO₃S
  • Storage temperature:
    2-8°C
  • Description:
    Methiocarb-sulfoxide
  • Form:
    Solid

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.