Pesticide standard, Aldicarb-sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1677-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:1646-88-4
- Description:Aldicarb-sulfone
- Formula:C₇H₁₄N₂O₄S
- Storage temperature:Ambient
- Description:Aldicarb-sulfone
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.