Pesticide standard, Pirimiphos-methyl, reference material, Honeywell Fluka™
HONCR1675-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:29232-93-7
- Description:Pirimiphos-methyl
- Formula:C₁₁H₂₀N₃O₃PS
- Storage temperature:Ambient
- Description:Pirimiphos-methyl
- Form:Liquid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.