Order Entry
Austria
ContactUsLinkComponent
 
Pesticide standard, Iprobenfos, reference material, Honeywell Fluka™
Catalog # HONCR1640-100MG
CAS Number:  
Pesticide standard, Iprobenfos, reference material, Honeywell Fluka™
Catalog # HONCR1640-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    26087-47-8
  • Description:
    Iprobenfos
  • Formula:
    C₁₃H₂₁O₃PS
  • Storage temperature:
    2-8°C
  • Description:
    Iprobenfos
  • Form:
    Liquid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.