Pesticide standard, Dimethoate, reference material, Honeywell Fluka™
HONCR1625-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:60-51-5
- Description:Dimethoate
- Formula:C₅H₁₂NO₃PS₂
- Storage temperature:2-8°C
- Description:Dimethoate
- Form:Solid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.