Pesticide standard, Difenoconazol, reference material, Honeywell Fluka™
HONCR1624-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:119446-68-3
- Description:Difenoconazol
- Formula:C₁₉H₁₇Cl₂N₃O₃
- Storage temperature:Ambient
- Description:Difenoconazol
- Form:Solid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.