Pesticide standard, Dichlorvos, reference material, Honeywell Fluka™
Catalog # HONCR1620-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:62-73-7
- Description:Dichlorvos
- Formula:C₄H₇Cl₂O₄P
- Storage temperature:-20°C
- Description:Dichlorvos
- Form:Liquid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.