Pesticide standard, Fipronil, reference material, Honeywell Fluka™
HONCR1602-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:120068-37-3
- Description:Fipronil
- Formula:C₁₂H₄Cl₂F₆N₄OS
- Storage temperature:Ambient
- Description:Fipronil
- Form:Solid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.