Order Entry
Austria
ContactUsLinkComponent
Pesticide standard, Fipronil carboxamide, reference material, Honeywell Fluka™
Pesticide standard, Fipronil carboxamide, reference material, Honeywell Fluka™
Catalog # HONCR1270-10MG
CAS Number:  
Pesticide standard, Fipronil carboxamide, reference material, Honeywell Fluka™
Catalog # HONCR1270-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    205650-69-7
  • Description:
    Fipronil carboxamide
  • Melting point:
    225…230 °C
  • Formula:
    C₉D₆H₅ClN₂O
  • Storage temperature:
    Ambient
  • Description:
    Fipronil carboxamide
  • Form:
    Neat

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.