ContactUsLinkComponent
Beverage refrigerators, FKU series
Beverage refrigerators, FKU series
Supplier:  Liebherr
Certificates
undefined
Beverage refrigerators, FKU series
Supplier:  Liebherr
Specifications
  • Catalog No:
  • 472-2181
  • 472-2182
  • Model:
  • FKU 1800
  • FKU 1805
  • Cat. No.:
  • 472-2181
  • 472-2182
  • Display:
  • Digital
  • Digital
  • Defrost:
  • automatic
  • automatic
  • Shelves/drawers:
  • 3 plastic-coated grid shelves
  • 3 plastic-coated grid shelves
  • Door type:
  • 1 solid swing door
  • 1 stainless steel swing door
  • Door lock:
  • Yes
  • Yes
  • Weight:
  • 34 kg
  • 34 kg
Specifications
Related Information
Product Information