Order Entry
Austria
ContactUsLinkComponent
N,O-Bis-(trimethylsilyl)-trifluoracetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSTFA/TMCS/TMSI) mixture 3:3:2 (v/v/v) silylating mixture III, LiChropur™ for GC derivatisation, Supelco®
Flammable
Corrosive
Irritant
N,O-Bis-(trimethylsilyl)-trifluoracetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSTFA/TMCS/TMSI) mixture 3:3:2 (v/v/v) silylating mixture III, LiChropur™ for GC derivatisation, Supelco®
Catalog # 85433-10ML
Supplier:  Merck
CAS Number:  
undefined
Flammable
Corrosive
Irritant
N,O-Bis-(trimethylsilyl)-trifluoracetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSTFA/TMCS/TMSI) mixture 3:3:2 (v/v/v) silylating mixture III, LiChropur™ for GC derivatisation, Supelco®
Catalog # 85433-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
ADR:8 (3),II
 :M-Clarity™ quality level = MQ100
Flash Pt:3 °C
UN:2924
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
 :25561-30-2