Order Entry
Austria
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck

Specification Test Results

  • Appearance (Colour):
    Faint Yellow to Yellow
  • Appearance (Form):
    Liquid
  • Refractive index N20/D:
    1,434 - 1,438
  • Proton NMR Spectrum:
    Conforms to structure

Specification Test Results

Frequently Bought Together