Order Entry
Austria
ContactUsLinkComponent
Test mixture 2 for apolar capillary columns according to Grob, for GC
Test mixture 2 for apolar capillary columns according to Grob, for GC
Catalog # 86501-10ML
Supplier:  Merck
CAS Number:  
Test mixture 2 for apolar capillary columns according to Grob, for GC
Catalog # 86501-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    ready-to-use
  • Pk:
    10 mL
  • Description:
    Test mixture 2 for apolar capillary columns according to Grob

Specifications

About this item

Mixture of D(−)-2,3-butanediol, decane, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylcaproic acid, methyl decanoate, methyl laurate, methyl undecanoate, 1-octanol and dodecane in hexane and methylene chloride

Storage temperature: 2 to 8 °C