0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Phthalates - PT, Proficiency Testing Material, Supelco®
Phthalates - PT, Proficiency Testing Material, Supelco®
Catalog # PE1412-2ML
Supplier:
Merck
CAS Number:
Expand image
Phthalates - PT, Proficiency Testing Material, Supelco®
Catalog # PE1412-2ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
2 mL
Description:
Phthalates - PT
Type:
Proficiency Testing Material
Specifications
About this item
Product Details
Product Details