Order Entry
Austria
ContactUsLinkComponent
 
Pesticides 2 - WP, proficiency testing material, Supelco®
Catalog # PE1201-2ML
Supplier:  Merck
CAS Number:  
Pesticides 2 - WP, proficiency testing material, Supelco®
Catalog # PE1201-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 mL for dilution to 1L
  • Pk:
    2 mL
  • Description:
    Pesticides 2 - WP
  • Type:
    Proficiency Testing Material

Specifications

About this item