Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, EPA TCL Base-Neutrals Mix
Supelco® Organic Reference Standard, EPA TCL Base-Neutrals Mix
Catalog # 47991-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EPA TCL Base-Neutrals Mix
Catalog # 47991-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each component in dichloromethane
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    EPA TCL Base-Neutrals mix
  • Environmentally Preferable:
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H315 - H319 - H331 - H336 - H350 - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P273 - P301 + P312 - P302 + P352 - P304 + P340 + P311 - P305 + P351 + P338 - P308 + P313