EPA 8100 PAH additional components mix, certified reference material, Supelco®
Catalog # 44694-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mug/mL in dichloromethane
- Pk:1
- Description:EPA 8100 PAH Additional Components Mix
- M-Clarity quality level:MQ100
- Type:Certified reference material