Order Entry
Austria
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, PCB standard solution 7
  36989-2ML
 :  Merck
 :  
Supelco® Organic Reference Standard, PCB standard solution 7
  36989-2ML
 :  Merck
 :  

 

  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • Description:
    PCB standard solution 7
  • Storage conditions:
    No temperature limit
  • Hazard class description:
    H225 - H304 - H315 - H336 - H373 - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    PCB No 28, 52, 101, 138, 153, 180, 209 solution
  • Flash point:
    4 °C - closed cup

 

 

  • GHS signal: GHS02, GHS08, GHS07, GHS09
 : P210 - P273 - P301 + P310 + P331 - P302 + P352 - P314