Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Phenols Mix 1, analytical standard, Supelco®
Phenols Mix 1, analytical standard, Supelco®
861253.
:
Merck
:
Phenols Mix 1, analytical standard, Supelco®
861253.
:
Merck
:
Concentration:
in isopropanol (varied conc.)
Pk:
1
Description:
Phenols Mix 1
Type:
Analytical standard