EPA 502/524 Volatiles Organic Calibration Mix (without gases), analytical standard, Supelco®
Catalog # 502111.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in methanol
- Pk:1
- Description:EPA 502/524 Volatiles Organic Calibration Mix (without gases)
- Type:Analytical standard