Order Entry
Austria
ContactUsLinkComponent
EPA TCL PAH Mix, certified reference material, Supelco®
EPA TCL PAH Mix, certified reference material, Supelco®
Catalog # 49156.
Supplier:  Merck
CAS Number:  
EPA TCL PAH Mix, certified reference material, Supelco®
Catalog # 49156.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in acetonitrile: methanol (9:1) (varied)
  • Pk:
    1
  • Description:
    EPA TCL PAH Mix
  • Type:
    Certified reference material

Specifications

About this item