EPA 8270 base-neutral surrogate spike mix, certified reference material, Supelco®
Catalog # 48925.
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mug/mL each component in dichloromethane
- Pk:1
- Description:EPA 8270 Base-Neutral Surrogate Spike Mix
- Type:Certified reference material