EPA 8270 nitrosamines mix, certified reference material, Supelco®
Catalog # 48489.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in methanol
- Pk:1
- Description:EPA 8270 Nitrosamines Mix
- Type:Certified reference material