Order Entry
Austria
ContactUsLinkComponent
MISA Group 23 chlorinated hydrocarbon mix, analytical standard, Supelco®
MISA Group 23 chlorinated hydrocarbon mix, analytical standard, Supelco®
Catalog # 48136.
Supplier:  Merck
CAS Number:  
MISA Group 23 chlorinated hydrocarbon mix, analytical standard, Supelco®
Catalog # 48136.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in dichloromethane
  • Pk:
    1
  • Description:
    MISA Group 23 Chlorinated Hydrocarbon Mix
  • Type:
    Analytical standard

Specifications

About this item