Order Entry
Austria
ContactUsLinkComponent
EPA VOC mix 6, analytical standard, Supelco®
EPA VOC mix 6, analytical standard, Supelco®
Catalog # 47408.
Supplier:  Merck
CAS Number:  
EPA VOC mix 6, analytical standard, Supelco®
Catalog # 47408.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in methanol
  • Pk:
    1
  • Description:
    EPA VOC Mix 6
  • Type:
    Analytical standard

Specifications

About this item