Order Entry
Austria
ContactUsLinkComponent
Semivolatile Acids Surrogate Spike Mix, certified reference material, Supelco®
Semivolatile Acids Surrogate Spike Mix, certified reference material, Supelco®
Catalog # 47261.
Supplier:  Merck
CAS Number:  
Semivolatile Acids Surrogate Spike Mix, certified reference material, Supelco®
Catalog # 47261.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mug/mL each component in methanol
  • Pk:
    1
  • Description:
    Semivolatile Acids Surrogate Spike Mix
  • Type:
    Certified reference material

Specifications

About this item