Order Entry
Austria
ContactUsLinkComponent
EPA 8040A phenol calibration mix, analytical standard, Supelco®
EPA 8040A phenol calibration mix, analytical standard, Supelco®
Catalog # 47899.
Supplier:  Merck
CAS Number:  
EPA 8040A phenol calibration mix, analytical standard, Supelco®
Catalog # 47899.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    500 mug/mL each component in isopropanol
  • Pk:
    1
  • Description:
    EPA 8040A Phenol Calibration Mix
  • Type:
    Analytical standard

Specifications

About this item