Supelco® Organic Reference Standard, 52 Component Indoor Air Standard
47537-U
: Merck
:
- Concentration:100 μg/ml each component in methanol: water (95:5)
- Pk:1
- Standards:Certified reference material
- Description:52 Component indoor air standard
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Japanese Indoor Air Standard Mix
- GHS signal: GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311