Supelco® Organic Reference Standard, Epa 506 Phthalate Mix (7 Component)
40077-U
: Merck
:
- Pk:1 mL
- Standards:Certified reference material, 1000 mug/ml each component in isooctane
- Description:EPA 506 Phthalate Mix (7 component)
- Storage conditions:2...30 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:–12 °C
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331