EPA 625 semivolatile calibration mix, analytical standard, Supelco®
Catalog # 506559.
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mug/mL each component in methylene chloride: benzene (3:1)
- Pk:1
- Description:EPA 625 Semivolatile Calibration Mix
- Type:Analytical standard