Order Entry
Austria
ContactUsLinkComponent
EPA 502 Internal Standard Mix, certified reference material, Supelco®
EPA 502 Internal Standard Mix, certified reference material, Supelco®
Catalog # 48950-U
Supplier:  Merck
CAS Number:  
EPA 502 Internal Standard Mix, certified reference material, Supelco®
Catalog # 48950-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in methanol
  • Pk:
    1
  • Description:
    EPA 502 Internal Standard Mix
  • M-Clarity quality level:
    MQ100
  • Type:
    Certified reference material

Specifications

About this item