Order Entry
Austria
ContactUsLinkComponent
EPA 610 polynuclear aromatic hydrocarbons mixture, certified reference material, Supelco®
EPA 610 polynuclear aromatic hydrocarbons mixture, certified reference material, Supelco®
Catalog # 4S8743
Supplier:  Merck
CAS Number:  
EPA 610 polynuclear aromatic hydrocarbons mixture, certified reference material, Supelco®
Catalog # 4S8743
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in methanol: methylene chloride (1:1) (varied)
  • Pk:
    1
  • Description:
    EPA 610 Polynuclear Aromatic Hydrocarbons Mixture
  • Type:
    Certified reference material

Specifications

About this item