Supelco® PESTANAL® Organic Reference Standard, Zoxamide
Catalog # 32501-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:156052-68-5
- Description:Zoxamide
- Storage conditions:2...30 °C
- Hazard class description:H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD03792722
- M-Clarity quality level:MQ100
- Synonym(s):3,5-Dichloro-N-(3-chloro-1-ethyl-1-methyl-2-oxopropyl)-4-methylbenzamide
- Formula:C₁₄H₁₆Cl₃NO₂
- Molecular weight:336,64
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P272 - P273 - P280 - P302 + P352 - P333 + P313