Supelco® PESTANAL® Organic Reference Standard, Triafamone
34552-50MG
: Merck
:
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:874195-61-6
- Description:Triafamone
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):2′-[(4,6-Dimethoxy-1,3,5-triazin-2-yl)carbonyl]-1,1,6′-trifluoro-N-methylmethanesulfonanilide; N-[2-[(4,6-Dimethoxy-1,3,5-triazin-2-yl)carbonyl]-6-fluorophenyl]-1,1-difluoro-N-methylmethanesulfonamide
- Formula:C₁₄H₁₃F₃N₄O₅S
- Molecular weight:406,34
- Flash point:Not applicable
- GHS signal: GHS09
: P273 - P391 - P501