Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1015855-87-4
- Description:Ronidazole-d3, VETRANAL(TM)
- M-Clarity quality level:MQ100
- Formula:C₆D₃H₅N₄O₄
- Description:Ronidazole-d3, VETRANAL(TM)
Ronidazole-d₃ may be used internal standard in the determination of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue using ultra-high performance liquid chromatography coupled with mass spectrometry (UHPLC/MS-MS).