Pesticide reference standard, Pyrithiobac, analytical standard, Supelco®
09163-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:123342-93-8
- Description:Pyrithiobac
- Melting point:157...163 °C
- Formula:C₁₃H₁₁ClN₂O₄S
- Description:Pyrithiobac
: Bottomless glass bottles are inserted with fused cone.