Order Entry
Austria
ContactUsLinkComponent
 
Pesticide reference standard, Promecarb, PESTANAL®, analytical standard, Supelco®
  45634-50MG
 :  Merck
 :  
undefined
Pesticide reference standard, Promecarb, PESTANAL®, analytical standard, Supelco®
  45634-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2631-37-0
  • Description:
    Promecarb
  • Formula:
    C₁₂H₁₇NO₂
  • Description:
    Promecarb

 

 

Promecarb may be used as a reference standard for the determination of promecarb in water by solid-phase extraction and liquid chromatography coupled with ionspray mass spectrometry.

  • Contains N-methylcarbamate group
  • Mode of action involves inhibiting the cholinesterase enzyme activity