Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Phenmedipham-Ethyl
Supelco® PESTANAL® Organic Reference Standard, Phenmedipham-Ethyl
Catalog # 37079-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Phenmedipham-Ethyl
Catalog # 37079-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    13684-44-1
  • Description:
    Phenmedipham-ethyl
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD13194954
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Ethoxycarbonylaminophenyl 3-methylcarbanilate; 3-[(Ethoxycarbonyl)amino]phenyl N-(3-methylphenyl)carbamate; Ethyl 3-(3-methylcarbaniloyloxy)carbanilate
  • Formula:
    C₁₇H₁₈N₂O₄
  • Molecular weight:
    314,34
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P391 - P501