Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Pesticide Standard 17 Solution
Supelco® PESTANAL® Organic Reference Standard, Pesticide Standard 17 Solution
Catalog # 36978-2ML
Supplier:  Merck
CAS Number:  
Supelco® PESTANAL® Organic Reference Standard, Pesticide Standard 17 Solution
Catalog # 36978-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Standards:
    PESTANAL®, Analytical standard, according to DIN 38407, in acetonitrile
  • Description:
    Pesticide standard 17 solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    2...8 °C
  • Flash point:
    2 °C

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338